4-(N,N-Diphenylamino)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Diphenylamino)benzeneboronic acid, 4-(Diphenylamino)phenylboronic acid
H52902-03
H52902-MD
AAH52902-03EA
165
USD
AAH52902-03
AAH52902-MD
4-(N,N-Diphenylamino)phenylboronic acid 98%
4-(N,N-Diphenylamino)phenylboronic acid
Purity: 98%
CAS No.: 201802-67-7
Molecular Formula: C18H16BNO2
Formula Weight: 289.14
Melting Point: 110-115°
MDL No.: MFCD06798117
Formula:
C₁₈H₁₆BNO₂ Melting Pt: 110…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798117 CAS Number: 201802-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...