5-Methyl-3-pyridineboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Methylpyridine-3-boronic acid
H52891-03
H52891-MD
AAH52891-03EA
95
USD
AAH52891-03
AAH52891-MD
5-Methyl-3-pyridineboronic acid 98%
5-Methyl-3-pyridineboronic acid
Purity: 98%
CAS No.: 173999-18-3
Molecular Formula: C6H8BNO2
Formula Weight: 136.95
Storage Temperature: -30°C to -10°C
Melting Point:
Formula:
C₆H₈BNO₂ MW: 136.95 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03428528 CAS Number: 173999-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...