4-Butoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Butoxyphenyl)boronic acid, 4-n-Butoxyphenylboronic acid

H52865-03 H52865-06
AAH52865-03EA 39 USD
AAH52865-03 AAH52865-06
4-Butoxyphenylboronic acid 98%
4-Butoxyphenylboronic acid

Purity: 98% CAS No.: 105365-51-3 Molecular Formula: C10H15BO3 Formula Weight: 194.04 Melting Point: 102-104° MDL No.: MFCD03427054

Formula: C₁₀H₁₅BO₃
MW: 194.04 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD03427054
CAS Number: 105365-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR