4-Methoxycarbonyl-2-nitrophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Methoxycarbonyl)-2-nitrophenylboronic acid, 4-(Methoxycarbonyl)-2-nitrobenzeneboronic acid, 4-Methoxycarbonyl-2-nitrobenzeneboronic acid
H52863-03
H52863-06
AAH52863-03EA
33.2
USD
AAH52863-03
AAH52863-06
4-Methoxycarbonyl-2-nitrophenylboronic acid 97%
4-Methoxycarbonyl-2-nitrophenylboronic acid
Purity: 97%
CAS No.: 85107-55-7
Molecular Formula: C8H8BNO6
Formula Weight: 224.96
Melting Point: 160-171°
MDL No.: MFCD01632202
Formula:
C₈H₈BNO₆ Melting Pt: 160…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632202 CAS Number: 85107-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...