5-Fluoro-3-pyridineboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H52860-03 H52860-MD
AAH52860-03EA 331 USD
AAH52860-03 AAH52860-MD
5-Fluoro-3-pyridineboronic acid 98%
5-Fluoro-3-pyridineboronic acid

Purity: 98% CAS No.: 872041-86-6 Molecular Formula: C5H5BFNO2 Formula Weight: 140.91 Melting Point: 233-236° MDL No.: MFCD07368243

Formula: C₅H₅BFNO₂
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD07368243
CAS Number: 872041-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR