(2-(Methoxymethyl)phenyl)boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-methoxymethylphenyl)boronic acid, 2-(Methoxymethyl)phenylboronic acid, 2-(Methoxymethyl)benzeneboronic acid, 2-Methoxymethylphenylboronic acid

H52858-03 H52858-MD
AAH52858-03EA 151 USD
AAH52858-03 AAH52858-MD
(2-(Methoxymethyl)phenyl)boronic acid 97%
(2-(Methoxymethyl)phenyl)boronic acid

Purity: 97% CAS No.: 126617-98-9 Molecular Formula: C8H11BO3 Formula Weight: 165.99 MDL No.: MFCD03412110

Formula: C₈H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD03412110
CAS Number: 126617-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR