4-(Aminomethyl)phenylboronic acid hydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52855-03
H52855-06
AAH52855-03EA
142
USD
AAH52855-03
AAH52855-06
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
4-(Aminomethyl)phenylboronic acid hydrochloride
Purity: 96%
CAS No.: 75705-21-4
Molecular Formula: C7H10BNO2.HCl
Formula Weight: 187.43
Melting Point: 240-250°
MDL No.: MFCD01632199
Formula:
C₇H₁₁BClNO₂ Melting Pt: 240…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632199 CAS Number: 75705-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...