4-Nitrophenylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester

H52848-03 H52848-06
AAH52848-03EA 34.1 USD
AAH52848-03 AAH52848-06
4-Nitrophenylboronic acid pinacol ester 98%
4-Nitrophenylboronic acid pinacol ester

Purity: 98% CAS No.: 171364-83-3 Molecular Formula: C12H16BNO4 Formula Weight: 249.08 Melting Point: 108-109° MDL No.: MFCD02179437

Formula: C₁₂H₁₆BNO₄
Melting Pt: 108…109 °C
Storage Temperature: Ambient
MDL Number: MFCD02179437
CAS Number: 171364-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR