N,N-Dimethyl 3-boronobenzenesulfonamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-(N,N-Dimethylsulphamoyl)phenyl)boronic acid, 3-(Dimethylsulphamoyl)benzeneboronic acid

H52838-03 H52838-MD
AAH52838-03EA 306 USD
AAH52838-03 AAH52838-MD
N,N-Dimethyl 3-boronobenzenesulfonamide 98%
N,N-Dimethyl 3-boronobenzenesulphonamide

Purity: 98% CAS No.: 871329-59-8 Molecular Formula: C8H12BNO4S Formula Weight: 229.06 Melting Point: 88-92° MDL No.: MFCD07363747

Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD07363747
CAS Number: 871329-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR