Boc-O-methyl-L-serine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-2-amino-3-methoxypropionic acid, Boc-Ser(Me)-OH, N-tert-Butoxycarbonyl-O-methyl-L-serine, (2S)-2-Amino-3-methoxypropanoic acid, N-BOC protected

H52829-03 H52829-MD
AAH52829-03EA 211 USD
AAH52829-03 AAH52829-MD
Boc-O-methyl-L-serine 97%
Boc-O-methyl-L-serine

Purity: 97% CAS No.: 51293-47-1 Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL No.: MFCD00153314

Formula: C₉H₁₇NO₅
MW: 219.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153314
CAS Number: 51293-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR