Fmoc-4-Abz-OH 97%

Supplier: Thermo Scientific Chemicals

H52828-03 H52828-MD
AAH52828-03EA 109 USD
AAH52828-03 AAH52828-06 AAH52828-MD
Fmoc-4-Abz-OH 97%
Fmoc-4-Abz-OH

Purity: 97% CAS No.: 185116-43-2 Molecular Formula: C22H17NO4 Formula Weight: 359.38 Beilstein Reference: 7722986 MDL No.: MFCD00144888

Formula: C₂₂H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00144888
CAS Number: 185116-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR