(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-D-Dap-OH

H52821-03 H52821-06 H52821-MD
AAH52821-03EA 155 USD
AAH52821-03 AAH52821-06 AAH52821-MD
(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid

Purity: 97% CAS No.: 76387-70-7 Molecular Formula: C8H16N2O4 Formula Weight: 204.23 Melting Point: 200-203° Optical Rotation: +14° (c=1 in water) MDL No.: MFCD01632072

Formula: C₈H₁₆N₂O₄
MW: 204.2 g/mol
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD01632072
CAS Number: 76387-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR