1-Boc-4-(aminomethyl)piperidine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

H52819-06 H52819-14
AAH52819-06EA 66.5 USD
AAH52819-06 AAH52819-14
1-Boc-4-(aminomethyl)piperidine 97%
1-Boc-4-(aminomethyl)piperidine

Purity: 97% CAS No.: 144222-22-0 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 Flash Point: 110°(230°F) Boiling Point: 237-238° MDL No.: MFCD01076207

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 237…238 °C
Density: 1.013
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01076207
CAS Number: 144222-22-0
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR