3-Amino-1-Boc-piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

H52816-03 H52816-06
AAH52816-03EA 58.3 USD
AAH52816-03 AAH52816-06
3-Amino-1-Boc-piperidine 97%
3-Amino-1-Boc-piperidine

Purity: 97% CAS No.: 184637-48-7 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD01861219

Formula: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01861219
CAS Number: 184637-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR