tert-Butyl-4-formylpiperidine-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 4-formylpiperidine-1-carboxylate, N-Boc-4-Formylpiperidine, 1-BOC-4-Piperidinecarboxaldehyde, 1-Boc-4-formylpiperidine, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxaldehyde, 1-(tert-Butoxycarbonyl)-4-formylpiperidine, 1-Boc-4-piperidinecarboxaldehyde, Tert-butyl 4-formylpiperidine-1-carboxylate

H52813-03 H52813-06
AAH52813-03EA 87.3 USD
AAH52813-03 AAH52813-06 AAH52813-MD
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
tert-Butyl-4-formylpiperidine-1-carboxylate

Purity: 97% CAS No.: 137076-22-3 Molecular Formula: C11H19NO3 Formula Weight: 213.28 MDL No.: MFCD02179019

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179019
CAS Number: 137076-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR