1-Boc-3-methylpiperazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl 3-methylpiperazine-1-carboxylate, 3-Methylpiperazine, N1-BOC protected
H52809-06
H52809-14
AAH52809-06EA
80.3
USD
AAH52809-06
AAH52809-14
1-Boc-3-methylpiperazine 97%
1-Boc-3-methylpiperazine
Purity: 97%
CAS No.: 120737-59-9
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
MDL No.: MFCD03001706
Formula:
C₁₀H₂₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03001706 CAS Number: 120737-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...