Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H52806-03 H52806-MD
AAH52806-03EA 733 USD
AAH52806-03 AAH52806-MD
Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid 97%
Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Purity: 97% CAS No.: 73397-28-1 Molecular Formula: C12H23NO5 Formula Weight: 261.32 MDL No.: MFCD04972275

Formula: C₁₂H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD04972275
CAS Number: 73397-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR