tert-Butyl 2-phenylpiperazine-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenylpiperazine, N1-BOC protected, 1-Boc-2-phenylpiperazine, 1-(tert-Butoxycarbonyl)-2-phenylpiperazine, 2-Methyl-2-propanyl 2-phenyl-1-piperazinecarboxylate

H52797-03 H52797-MD
AAH52797-03EA 333 USD
AAH52797-03 AAH52797-MD
tert-Butyl 2-phenylpiperazine-1-carboxylate 97%
tert-Butyl 2-phenylpiperazine-1-carboxylate

Purity: 97% CAS No.: 886766-60-5 Molecular Formula: C15H22N2O2 Formula Weight: 262.35 MDL No.: MFCD07367734

Formula: C₁₅H₂₂N₂O₂
MW: 262.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367734
CAS Number: 886766-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR