Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide
H52795-03
H52795-06
H52795-MD
AAH52795-03EA
306
USD
AAH52795-03
AAH52795-06
AAH52795-MD
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) 97%
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)
Purity: 97%
CAS No.: 106627-54-7
Molecular Formula: C4H4NNaO6S
Formula Weight: 217.13
Melting Point: 250°
Beilstein Reference: 6359129
MDL No.: MFCD00043100
Formula:
C₄H₄NNaO₆S MW: 217.13 g/mol Melting Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043100 CAS Number: 106627-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...