1-Boc-azetidine-3-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Formylazetidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde, tert-Butyl 3-Formylazetidine-1-carboxylate, 1-Boc-azetidine-3-carboxaldehyde
H52794-03
H52794-06
H52794-MD
AAH52794-03EA
156
USD
AAH52794-03
AAH52794-06
AAH52794-MD
1-Boc-azetidine-3-carboxaldehyde 97%
1-Boc-azetidine-3-carboxaldehyde
Purity: 97%
CAS No.: 177947-96-5
Molecular Formula: C9H15NO3
Formula Weight: 185.22
Storage Temperature: 2°C to 8°C
Boiling Point: 77-82°/2mm
MDL No.: MFCD06656142
Formula:
C₉H₁₅NO₃ MW: 185.22 g/mol Boiling Pt: 77…82 °C/2 mm Storage Temperature: Ambient |
MDL Number:
MFCD06656142 CAS Number: 177947-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...