Boc-(S)-thiazolidine-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-3-(tert-Butoxycarbonyl)thiazolidine-2-carboxylic acid

H52793-03 H52793-06
AAH52793-03EA 128 USD
AAH52793-03 AAH52793-06 AAH52793-MD
Boc-(S)-thiazolidine-2-carboxylic acid 97%
Boc-(S)-thiazolidine-2-carboxylic acid

Purity: 97% CAS No.: 891192-95-3 Molecular Formula: C9H15NO4S Formula Weight: 233.28 Melting Point: 90-93° Optical Rotation: -47° (c=5 in methanol) MDL No.: MFCD02682342

Formula: C₉H₁₅NO₄S
MW: 233.29 g/mol
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD02682342
CAS Number: 891192-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR