Boc-D-α-methylserine 97%

Supplier: Thermo Scientific Chemicals

H52787-03 H52787-MD
AAH52787-03EA 684 USD
AAH52787-03 AAH52787-MD
Boc-D-α-methylserine 97%
Boc-D-alpha-methylserine

Purity: 97% CAS No.: 84311-18-2 Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL No.: MFCD02682596

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682596
CAS Number: 84311-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR