cis-N-Boc-4-cyano-L-proline methyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-cis-4-CN-Pro-OMe, (2S,4S)-1-tert-Butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, 1-tert-Butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate

H52783-03 H52783-MD
AAH52783-03EA 573 USD
AAH52783-03 AAH52783-MD
cis-N-Boc-4-cyano-L-proline methyl ester 97%
cis-N-Boc-4-cyano-L-proline methyl ester

Purity: 97% CAS No.: 487048-28-2 Molecular Formula: C12H18N2O4 Formula Weight: 254.29 MDL No.: MFCD03787928

Formula: C₁₂H₁₈N₂O₄
MW: 254.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787928
CAS Number: 487048-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR