Boc-(S)-2-amino-3-ethoxypropionic acid 97%
Supplier: Thermo Scientific Chemicals
H52781-03
H52781-MD
AAH52781-03EA
484
USD
AAH52781-03
AAH52781-MD
Boc-(S)-2-amino-3-ethoxypropionic acid 97%
Boc-(S)-2-amino-3-ethoxypropionic acid
Purity: 97%
CAS No.: 104839-00-1
Molecular Formula: C10H19NO5
Formula Weight: 233.26
MDL No.: MFCD06797558
Formula:
C₁₀H₁₉NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD06797558 CAS Number: 104839-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...