(1S,2S)-2-Aminocyclohexanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H52779-03 H52779-06 H52779-MD
AAH52779-03EA 321 USD
AAH52779-03 AAH52779-06 AAH52779-MD
(1S,2S)-2-Aminocyclohexanecarboxylic acid 97%
(1S,2S)-2-Aminocyclohexanecarboxylic acid

Purity: 97% CAS No.: 24716-93-6 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Optical Rotation: +77° (c=1 in water) MDL No.: MFCD01318270

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318270
CAS Number: 24716-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR