4-Amino-2-bromopyrimidine-5-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H52776-03 H52776-06
AAH52776-03EA 83.6 USD
AAH52776-03 AAH52776-06 AAH52776-MD
4-Amino-2-bromopyrimidine-5-carbonitrile 97%
4-Amino-2-bromopyrimidine-5-carbonitrile

Purity: 97% CAS No.: 94741-70-5 Molecular Formula: C5H3BrN4 Formula Weight: 199.01 Melting Point: 254° dec. MDL No.: MFCD00173661

Formula: C₅H₃BrN₄
MW: 199.01 g/mol
Melting Pt: 254 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00173661
CAS Number: 94741-70-5
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR