1-Boc-3-(methylamino)piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine
H52773-03
H52773-06
AAH52773-03EA
47
USD
AAH52773-03
AAH52773-06
1-Boc-3-(methylamino)piperidine 97%
1-Boc-3-(methylamino)piperidine
Purity: 97%
CAS No.: 392331-89-4
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
Boiling Point: 95-99°/0.6mm
MDL No.: MFCD04114980
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol Boiling Pt: 95…99 °C/0.6 mm Storage Temperature: Ambient |
MDL Number:
MFCD04114980 CAS Number: 392331-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...