(S)-1-Boc-3-hydroxypiperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected

H52771-03 H52771-06
AAH52771-03EA 56.3 USD
AAH52771-03 AAH52771-06
(S)-1-Boc-3-hydroxypiperidine 97%
(S)-1-Boc-3-hydroxypiperidine

Purity: 97% CAS No.: 143900-44-1 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Flash Point: 110°(230°F) Physical Form: Powder Melting Point: 46-50° Optical Rotation: +23° (c=5 in methanol) MDL No.: MFCD04115307

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR