2-(Dimethylcarbamoyl)phenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Dimethylcarbamoyl)benzeneboronic acid

H52765-03 H52765-MD
AAH52765-03EA 229 USD
AAH52765-03 AAH52765-MD
2-(Dimethylcarbamoyl)phenylboronic acid 95%
2-(Dimethylcarbamoyl)phenylboronic acid

Purity: 95% CAS No.: 874219-16-6 Molecular Formula: C9H12BNO3 Formula Weight: 193.01 Melting Point: 108-110° MDL No.: MFCD03425959

Formula: C₉H₁₂BNO₃
MW: 193.01
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD03425959
CAS Number: 874219-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR