(R)-3-(Boc-amino)piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl N-[(3R)-piperidin-3-yl]carbamate, (R)-3-Boc-aminopiperidine, (R)-tert-Butyl piperidin-3-ylcarbamate

H52749-03 H52749-06 H52749-14
AAH52749-03EA 53.8 USD
AAH52749-03 AAH52749-06 AAH52749-14
(R)-3-(Boc-amino)piperidine 97%
(R)-3-(Boc-amino)piperidine

Purity: 97% CAS No.: 309956-78-3 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD03093382

Formula: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03093382
CAS Number: 309956-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR