Methyl-2,2-difluoro-2-(fluorosulfonyl)acetate 97%

Supplier: Thermo Scientific Chemicals
Danger

H52745-06 H52745-14
AAH52745-06EA 58 USD
AAH52745-06 AAH52745-14
Methyl-2,2-difluoro-2-(fluorosulfonyl)acetate 97%
Methyl 2,2-difluoro-2-(fluorosulphonyl)acetate

Purity: 97% CAS No.: 680-15-9 Molecular Formula: C3H3F3O4S Formula Weight: 192.11 Flash Point: 47°(117°F) Refractive Index: 1.351 Boiling Point: 117-118° Beilstein Reference: 1812896 UN No.: 2920 MDL No.: MFCD00144316

Formula: C₃H₃F₃O₄S
MW: 192.11
Boiling Pt: 117…118 °C
Density: 1.509
Flash Pt: 47 °C (117 °F)
Storage Temperature: Ambient
MDL Number: MFCD00144316
CAS Number: 680-15-9
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR