Fmoc-Hyp-OH 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-trans-4-hydroxy-L-proline

H52740-03 H52740-06
AAH52740-03EA 28.7 USD
AAH52740-03 AAH52740-06
Fmoc-Hyp-OH 97%
Fmoc-Hyp-OH

Purity: 97% CAS No.: 88050-17-3 Molecular Formula: C20H19NO5 Formula Weight: 353.37 Melting Point: 189-193° Optical Rotation: -60° (c=1 in methanol) MDL No.: MFCD00151929

Formula: C₂₀H₁₉NO₅
MW: 353.374 g/mol
Melting Pt: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00151929
CAS Number: 88050-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR