(S)-1-Boc-2-(aminomethyl)pyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Danger

H52727-03 H52727-06
AAH52727-03EA 75.9 USD
AAH52727-03 AAH52727-06
(S)-1-Boc-2-(aminomethyl)pyrrolidine 97%
(S)-1-Boc-2-(aminomethyl)pyrrolidine

Purity: 97% CAS No.: 119020-01-8 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Boiling Point: 98-112°/1mm UN No.: 2735 MDL No.: MFCD03419257

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 98…112 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD03419257
CAS Number: 119020-01-8
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR