N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine 97%
Supplier: Thermo Scientific Chemicals
H52726-03
H52726-06
AAH52726-03EA
144
USD
AAH52726-03
AAH52726-06
AAH52726-MD
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine 97%
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine
Purity: 97%
Molecular Formula: C20H33N5O5Si
Formula Weight: 451.60
MDL No.: MFCD04972287
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...