tert-Butyl (piperidin-4-ylmethyl)carbamate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (4-Piperidylmethyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Piperidylmethyl)carbamate, 4-(tert-Butoxycarbonylaminomethyl)piperidine, 4-(Boc-aminomethyl)piperidine

H52725-03 H52725-06
AAH52725-03EA 65.4 USD
AAH52725-03 AAH52725-06
tert-Butyl (piperidin-4-ylmethyl)carbamate 97%
tert-Butyl (piperidin-4-ylmethyl)carbamate

Purity: 97% CAS No.: 135632-53-0 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 Melting Point: 106-108° UN No.: 3259 MDL No.: MFCD01631214

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01631214
CAS Number: 135632-53-0
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR