4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine hydrochloride, 4-Aminomethylphenylboronic acid pinacol ester HCl

H52691-03 H52691-MD
AAH52691-03EA 135 USD
AAH52691-03 AAH52691-MD
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride 95%
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride

Purity: 95% CAS No.: 850568-55-7 Molecular Formula: C13H20BNO2.HCl Formula Weight: 269.58 Melting Point: 228-238° MDL No.: MFCD02179455

Formula: C₁₃H₂₀BNO₂·HCl
MW: 269.58 g/mol
Melting Pt: 228…238 °C
Storage Temperature: Ambient
MDL Number: MFCD02179455
CAS Number: 850568-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR