4-Isopropoxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Isopropoxybenzeneboronic acid
H52675-03
H52675-06
H52675-14
AAH52675-03EA
28.7
USD
AAH52675-03
AAH52675-06
AAH52675-14
4-Isopropoxyphenylboronic acid 97%
4-Isopropoxyphenylboronic acid
Purity: 97%
CAS No.: 153624-46-5
Molecular Formula: C9H13BO3
Formula Weight: 180.01
Melting Point: 150-154°
MDL No.: MFCD03427051
Formula:
C₉H₁₃BO₃ MW: 180.01 g/mol Melting Pt: 150…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427051 CAS Number: 153624-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...