1-Boc-5-methoxyindole-2-boronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid
H52654-03
H52654-06
AAH52654-03EA
46.2
USD
AAH52654-03
AAH52654-06
1-Boc-5-methoxyindole-2-boronic acid 95%
1-Boc-5-methoxyindole-2-boronic acid
Purity: 95%
CAS No.: 290331-71-4
Molecular Formula: C14H18BNO5
Formula Weight: 291.11
Storage Temperature: -30°C to -10°C
Melting Point: 105-108°
MDL No.: MFCD04039006
Formula:
C₁₄H₁₈BNO₅ MW: 291.11 Melting Pt: 105…108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039006 CAS Number: 290331-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...