4-Pyridineboronic acid hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52648-03
H52648-06
AAH52648-03EA
45.3
USD
AAH52648-03
AAH52648-06
4-Pyridineboronic acid hydrochloride 95%
4-Pyridineboronic acid hydrochloride
Purity: 95%
CAS No.: 913835-65-1
Molecular Formula: C5H6BNO2.HCl
Formula Weight: 159.38
Storage Temperature: 2°C to 8°C
Melting Point: 340°
MDL No.: MFCD06201888
Formula:
C₅H₇BClNO₂ Melting Pt: 340 °C Storage Temperature: Ambient |
MDL Number:
MFCD06201888 CAS Number: 913835-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...