(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-2-(hydroxymethyl)benzeneboronic acid dehydrate hydrochloride, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol hydrochloride
H52629-03
H52629-MD
AAH52629-03EA
630
USD
AAH52629-03
AAH52629-MD
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 95%
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride
Purity: 95%
CAS No.: 117098-93-8
Molecular Formula: C7H8BNO2.HCl
Formula Weight: 185.42
Melting Point: 300°
MDL No.: MFCD04115645
Formula:
C₇H₉BClNO₂ MW: 185.42 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115645 CAS Number: 117098-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...