4-(Acetamidomethyl)benzeneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-[(Acetylamino)methyl]benzeneboronic acid, (4-(Acetamidomethyl)phenyl)boronic acid

H52602-03 H52602-MD
AAH52602-03EA 253 USD
AAH52602-03 AAH52602-MD
4-(Acetamidomethyl)benzeneboronic acid 97%
4-(Acetamidomethyl)benzeneboronic acid

Purity: 97% CAS No.: 850568-41-1 Molecular Formula: C9H12BNO3 Formula Weight: 193.01 Melting Point: 278-280° MDL No.: MFCD06659818

Formula: C₉H₁₂BNO₃
Melting Pt: 278…280 °C
Storage Temperature: Ambient
MDL Number: MFCD06659818
CAS Number: 850568-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR