3-Aminophenylboronic acid hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Aminophenyl)boronic acid hydrochloride, 3-Aminobenzeneboronic acid hydrochloride

H52595-03 H52595-06
AAH52595-03EA 59.8 USD
AAH52595-03 AAH52595-06
3-Aminophenylboronic acid hydrochloride 98%
3-Aminophenylboronic acid hydrochloride

Purity: 98% CAS No.: 85006-23-1 Molecular Formula: C6H8BNO2.HCl Formula Weight: 173.41 Melting Point: 300° MDL No.: MFCD00191748

Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191748
CAS Number: 85006-23-1
EINECS: 285-052-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR