(S)-1-Boc-piperidine-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52575-06
H52575-14
AAH52575-06EA
98.8
USD
AAH52575-06
AAH52575-14
(S)-1-Boc-piperidine-3-carboxylic acid 97%
(S)-1-Boc-piperidine-3-carboxylic acid
Purity: 97%
CAS No.: 88495-54-9
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Melting Point: 165-169°
Optical Rotation: +57° (c=1 in chloroform)
Formula:
C₁₁H₁₉NO₄ Melting Pt: 165…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179172 CAS Number: 88495-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...