3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 5,6-dihydropyridine-1(2H)-carboxylate, 1-Boc-1,2,3,6-tetrahydropyridine, tert-Butyl 1,2,3,6-Tetrahydropyridine-1-carboxylate, 1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1,2,3,6-Tetrahydropyridine-1-carboxylic acid tert-Butyl ester

H52574-03 H52574-06
AAH52574-03EA 78.8 USD
AAH52574-03 AAH52574-06 AAH52574-MD
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester

Purity: 97% CAS No.: 85838-94-4 Molecular Formula: C10H17NO2 Formula Weight: 183.25 Boiling Point: 78-84°/5mm MDL No.: MFCD04972245

Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
Boiling Pt: 78…84 °C/5 mm
Density: 1.029
Storage Temperature: Ambient
MDL Number: MFCD04972245
CAS Number: 85838-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR