Boc-L-α-methylserine 97%

Supplier: Thermo Scientific Chemicals

H52570-03 H52570-MD
AAH52570-03EA 377.31 USD
AAH52570-03 AAH52570-MD
Boc-L-α-methylserine 97%
Boc-L-alpha-methylserine

Purity: 97% CAS No.: 84311-19-3 Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL No.: MFCD02682593

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682593
CAS Number: 84311-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR