(S)-2-Amino-3-methoxypropanoic acid hydrochloride 97%
Supplier: Thermo Scientific Chemicals
H52562-03
H52562-MD
AAH52562-03EA
443
USD
AAH52562-03
AAH52562-MD
(S)-2-Amino-3-methoxypropanoic acid hydrochloride 97%
(S)-2-Amino-3-methoxypropanoic acid hydrochloride
Purity: 97%
CAS No.: 336100-47-1
Molecular Formula: C4H9NO3.HCl
Formula Weight: 155.58
Melting Point: 182-183°
MDL No.: MFCD03840366
Formula:
C₄H₁₀ClNO₃ MW: 155.58 g/mol Melting Pt: 182…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840366 CAS Number: 336100-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...