(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-1-BOC-3-(hydroxymethyl)piperidine, (R)-tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(R)-N-BOC-PIPERIDINE METHANOL, 1-Piperidinecarboxylicacid, 3-(hydroxymethyl)-, 1,1-dimethylethyl ester, (3R)-
H52559-03
H52559-06
AAH52559-03EA
48.6
USD
AAH52559-03
AAH52559-06
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate
Purity: 97%
CAS No.: 140695-85-8
Molecular Formula: C11H21NO3
Formula Weight: 215.29
Melting Point: 91-94°
Optical Rotation: -20° (c=2 in methanol)
MDL No.: MFCD02683202
Formula:
C₁₁H₂₁NO₃ MW: 215.293 g/mol Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683202 CAS Number: 140695-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...