2-Bromoimidazole 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H52553-03
H52553-06
AAH52553-03EA
212
USD
AAH52553-03
AAH52553-06
2-Bromoimidazole 95%
2-Bromoimidazole
Purity: 95%
CAS No.: 16681-56-4
Molecular Formula: C3H3BrN2
Formula Weight: 146.98
Storage Temperature: 2°C to 8°C
Melting Point: 197°
MDL No.: MFCD02179526
Formula:
C₃H₃BrN₂ Melting Pt: 197 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179526 CAS Number: 16681-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...