3-(2-Methoxycarbonylethyl)benzeneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-(3-Methoxy-3-oxopropyl)phenyl)boronic acid

H52542-03 H52542-MD
AAH52542-03EA 249 USD
AAH52542-03 AAH52542-MD
3-(2-Methoxycarbonylethyl)benzeneboronic acid 97%
3-(2-Methoxycarbonylethyl)benzeneboronic acid

Purity: 97% CAS No.: 833472-82-5 Molecular Formula: C10H13BO4 Formula Weight: 208.02 Melting Point: 96-100° MDL No.: MFCD04115648

Formula: C₁₀H₁₃BO₄
MW: 208.02 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD04115648
CAS Number: 833472-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR