1-Boc-6-cyanoindole-2-boronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Cyano-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-cyano-1H-indol-2-yl)boronic acid

H52520-03 H52520-MD
AAH52520-03EA 158 USD
AAH52520-03 AAH52520-MD
1-Boc-6-cyanoindole-2-boronic acid 96%
1-Boc-6-cyanoindole-2-boronic acid

Purity: 96% CAS No.: 913835-67-3 Molecular Formula: C14H15BN2O4 Formula Weight: 286.09 Storage Temperature: -30°C to -10°C Melting Point: 219-221° MDL No.: MFCD08436056

Formula: C₁₄H₁₅BN₂O₄
MW: 286.09 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436056
CAS Number: 913835-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR